4-Ethylanisol ≥98.0% (by GC)
Glass bottle, 25 mL
TCIAE0436-25ML
: TCI
: 1-Ethyl-4-methoxybenzene
- :1515-95-3
- :C₉H₁₂O
- :MFCD00015176
- :136.19 g/mol
- :195°C
- :0,96 g/cm³ (20 °C)
- :79°C
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Pakning:Glass bottle
- Pakn:25 mL



