Orders LinkContactUsLinkComponent
Non-aqueous DPX mounting medium
DPX new, Glass bottle, 500 ml
Katalog # 1.00579.0500
Leverandør:   Merck
Non-aqueous DPX mounting medium
Katalog # 1.00579.0500
Leverandør:  Merck
CAS nummer:  
Non-aqueous DPX mounting medium
DPX new, Glass bottle, 500 ml
CAS nummer:  
Price
644,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
DPX new is a synthetic non aqueous mounting medium for microscopy. In its formulation, the teratogenic plasticiizer dibutyl phthalate (DBP) has been avoided, which renders the composition safer for the environment and more user friendly than the conventionally used mounting agents such as DPX.
Preparative capacity (500 ml): 2500 slides
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle
  • Pakn:
    500 ml
  • Beskrivelse:
    DPX new
Specifikationer