Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Congo red ≥98.0% (by HPLC)

Congo red ≥98.0% (by HPLC)

Glass bottle, 25 g
Katalog # TCIAC0550-25G
Sundhed
Leverandør:   TCI
Congo red ≥98.0% (by HPLC)
Katalog # TCIAC0550-25G
Leverandør:  TCI
CAS nummer:  573-58-0
Congo red ≥98.0% (by HPLC)
Congo red ≥98.0% (by HPLC)

Glass bottle, 25 g

CAS nummer:  573-58-0
Price
706,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  CI 22120, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalen-1-sulfonat)
  • CAS nummer:573-58-0
  • Formel:C₃₂H₂₂N₆Na₂O₆S₂
  • MDL nummer:MFCD00004028
  • Molecular Weight:696.67 g/mol
  • Merck Index:14,02498
  • Smeltende Pt:> 360 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-358-4
Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark red to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Absorbance:
    min. 0.270 (10 mg/L, pH 5.0, 488.0 to 492.0 nm)
  • Solubility in hot Water:
    transparency
  • Drying loss:
    max. 8.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.