Lecithin (from soy bean)
Supplier: TCI
About this item
- CAS Number:8002-43-5
- MDL Number:MFCD00147406
- Merck Index:14,05429
- EINECS:232-307-2
2 Options Available Below
Product Details & Documents
- CAS Number:8002-43-5
- MDL Number:MFCD00147406
- Merck Index:14,05429
- EINECS:232-307-2
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to Amber to Dark purple clear liquid to slightly cloudy liquid
- Saponification Value:180.0 to 195.0
- Acetone insoluble matter:58.0 to 65.0 %
- Acid Value:max. 25.0