Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pentadecylcyclohexan ≥98.0% (by GC)

Pentadecylcyclohexan ≥98.0% (by GC)

Leverandør:   TCI
Pentadecylcyclohexan ≥98.0% (by GC)
Leverandør:  TCI
Certifikater
Pentadecylcyclohexan ≥98.0% (by GC)

2 tilgængelige muligheder nedenfor

Product Details & Documents
  • CAS nummer:6006-95-7
  • Formel:C₂₁H₄₂
  • MDL nummer:MFCD00058952
  • Molecular Weight:294.56 g/mol
  • Kogende Pt:373 °C (1013 hPa)
  • Smeltende Pt:29 °C
  • Tæthed:1,03 g/cm³ (0 °C)
  • EINECS:227-858-0
Specifications for Product Family

Specifikationstestresultater

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Freezing point:
    24.0 to 26.0 deg-C
Product Family Options

Produktinformation

  • Pakning
    Pakn
    Tilgængelighed
    Price
  • Glass bottle
    5 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 g
    1 * 5 g - 1.520,00 kr
    1.520,00 kr
    1 * 5 g
       1.520,00 kr

  • Glass bottle
    25 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 g
    1 * 25 g - 5.910,00 kr
    5.910,00 kr
    1 * 25 g
       5.910,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.