Natriumacetat
Glass bottle for solids, 1 kg
Katalog # RC-090
Leverandør: G-Biosciences
Gå til:
Product Details & Documents
Synonymer: Acetic acid sodium salt
- CAS nummer:127-09-3
- Formel:H₃CCOONa
- MDL nummer:MFCD00012459
- Molecular Weight:82.03 g/mol
- Merck Index:12,08711
- Kogende Pt:881,4 °C (1013 hPa)
- Smeltende Pt:324 °C
- Tæthed:1,528 g/cm³ (20 °C)
- Flash Pt:>250 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-823-8
- NÅ:01-2119485123-42
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg