Opsamlings- og analyseplader, Whatman UNIPLATE™
Catalog # 734-1217
Supplier: Whatman products (Cytiva)
Specifications
- Pakket:Individually wrapped
- Farve:ufarvet
- Beskrivelse:UNIPLATE, 24-well, 10 ml, round bottom, irradiated with lid
- Pakn:25
Specifications
About this item
PP, with clear PS lid. The Whatman™ UNIPLATE collection and analysis microplates accomodate 24 well plates, have round bottom wells.
- Chemical compatibility: Available in chemically resistant polymers capable of withstanding low temperatures for long-term storage. Opaque plates prevent optical cross talk in light emitting assays
- Guaranteed for use with robotic handlers and centrifuge carriers
Certifications: UNIPLATE microplates conform to the ANSI/SBS microplate standard and fit most microplate readers and automated plate handling devices.