Orders LinkContactUsLinkComponent
Dissolved oxygen electrodes
Dissolved oxygen electrodes
 :   KNICK
Dissolved oxygen electrodes
 :  KNICK
Dissolved oxygen electrodes
Dissolved oxygen electrodes
Product Family Options
  • Beskrivelse
    Model
    Koncentration
    Temperatur
    Materiale
    Tilslutning
    Ø×L
    Til
  • SE 340, optical oxygen probe, digital, with NTC 22 kOhm temperature sensor
    SE 340, optical, digital, with temperature sensor
    0 - 20 mg/l
    0...+50 °C
    POM
    M12, 8-pin, 1,5 m cable
    12×140 mm
    Water, wastewater
    1 * 1 Items
    1 * 1 Items - 12.290,00 kr
    12.290,00 kr
    1 * 1 Items
       12.290,00 kr
  • SE 715-MS, galvanic oxygen probe, digital, with NTC 22 kOhm temperature sensor
    SE 715/1-MS, digital, with temperature sensor
    0 - 20 mg/l
    −5...+45 °C
    Polysulfon
    Memosens
    12×120 mm
    Water, wastewater, aquariums
    1 * 1 Items
    1 * 1 Items - 12.280,00 kr
    12.280,00 kr
    1 * 1 Items
       12.280,00 kr