Orders LinkContactUsLinkComponent
Universal couplings, BOLA
Universal couplings, BOLA
Leverandør:   Bohlender
Universal couplings, BOLA
Leverandør:  Bohlender
Certifikater
Universal couplings, BOLA
Universal couplings, BOLA
2 tilgængelige muligheder nedenfor
Product Details & Documents
PTFE, white.
  • Temperature resistance: from –200 to +250 °C
  • Universal chemical resistance
  • FDA conform
Coupling for transition from M6 thread to UNF ¼" 28 G thread. The M6 thread is marked by a groove.
Product Family Options
Produktinformation
  • Gevind
    Materiale
    Tilgængelighed
    Price
  • BOHLF772-08
    outer thread M 6
    outer thread M 6
    male, UNF ¼" 28 G, outer thread
    PTFE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 535,00 kr
    535,00 kr
    1 * 1 Items
       535,00 kr
  • BOHLF770-08
    internal thread M 6
    internal thread M 6
    female, UNF ¼" 28 G, outer thread
    PTFE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 479,00 kr
    479,00 kr
    1 * 1 Items
       479,00 kr