PIG® HazMat chemical absorbent dikes
NEWPHA1010
: NEW PIG
- Beskrivelse:Chemical absorbent dikes
- Sugeevne:68 L
- Pakket:2 socks/box
- Pakn:2
- Ø×L:130×3000 mm
Your biggest spills require the stopping power of these large-diameter hazmat dikes.
- Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
- Chemical-resistant sock won't degrade or cause a dangerous reaction upon contact with corrosive spills
- Excellent for quickly diking and stopping the spread of larger chemical spills; for diking smaller chemical spills, consider our smaller diameter PIG HazMat Socks
- PP skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated
- PP filler is highly absorbent for containing corrosive or reactive spills
The pillow absorbs and retains most acids, caustics and unknown liquids. The pink colour is easily distinguishable to assure workers use the correct pillow during a spill emergency. Can be incinerated after use to reduce waste.
: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.