Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
100 ml, 29/32 NS
Katalog # IKAA1905700
Leverandør:   IKA
Evaporating flasks
Katalog # IKAA1905700
Leverandør:  IKA
Evaporating flasks
100 ml, 29/32 NS
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard IKA® rotary evaporators; with white labelling area.
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    100 ml
  • Seq:
    01