Microcentrifuge tubes
Katalog # KIMA18166
Leverandør: KIMA SAS DI CHIARIN RENZO
Specifikationer
- Model:with attached snap cap, Eppendorf type
- Version:graduated, conical bottom, non sterile
- Kapacitet:1,5 ml
- Pakn:1.000
Specifikationer
Om denne vare
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Non sterile
With attached push-on cap.