Ground joint reducing set, BOLA
Reducing set, NS 14, NS 19, NS 24, NS 29, NS 34, NS 45 and NS 60
BOHLH981-14
Ground joint reducing set, PTFE, consisting of reducing rings graduated for bridging different cone and socket sizes.
- Universal chemical resistance
- Temperature resistance –200 to +250 °C
- Suitable for vacuum applications
- Til:NS 14, NS 19, NS 24, NS 29, NS 34, NS 45 and NS 60
- Beskrivelse:Reducing set