Capillary columns, Elite-Guard and Elite-Siltek® for autosystem
PERCN9316608
- Kolonne form:Kapillær
- Ø int.:0,32 mm
- Beskrivelse:Elite-Siltek Guard
- Length:5 m
- Seq:08
- Environmentally Preferable:
- Pakn:1
These guard columns lengthen the life of the capillary column and improve the analyte focusing.
- Increase the life of the capillary column and performance
- Maintains retention time and resolution on capillary column
- Traps non-volatile particles
Both the intermediate-polarity deactivated guard columns and the Siltek® deactivated guard columns are important to match the polarity of the solvent and the polarity of the surface deactivation. Temperature range is 325 °C for Elite-Guardand 380 °C for Elite-Siltek® guard.