Orders LinkContactUsLinkComponent
PIG® medium and large mobile container kits
PIG® medium and large mobile container kits
 :   NEW PIG
PIG® medium and large mobile container kits
 :  NEW PIG
PIG® medium and large mobile container kits
PIG® medium and large mobile container kits
Fast, effective response to indoor and outdoor spills.
  • Durable, wheeled 240 and 360 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface
Weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Product Family Options
Medium mobile kits: 240 litres
  • Beskrivelse
    Sugeevne
    B×D×H
  • Spill kit, Universal, includes 150 mat pads, 16 socks, 12 pillows and 10 disposal bags
    183,1 L
    730×570×1050 mm
    1 * 1 Each
    1 * 1 Each - 4.250,00 kr
    4.250,00 kr
    1 * 1 Each
       4.250,00 kr
  • Refill only, Universal
    183,1 L
    1 * 1 Items
    1 * 1 Items - 2.450,00 kr
    2.450,00 kr
    1 * 1 Items
       2.450,00 kr
  • Spill kit, HAZ-MAT, includes 120 mat pads, 12 socks, 18 pillows and 10 disposal bags
    179,2 L
    730×570×1050 mm
    1 * 1 Each
    1 * 1 Each - 7.560,00 kr
    7.560,00 kr
    1 * 1 Each
       7.560,00 kr
  • Refill only, HAZ-MAT
    179,2 L
    1 * 1 Items
    1 * 1 Items - 3.970,00 kr
    3.970,00 kr
    1 * 1 Items
       3.970,00 kr
  • Spill kit, Oil-Only, includes 120 mat pads, 8 socks, 12 pillows and 10 disposal bags
    168,3 L
    730×570×1050 mm
    1 * 1 Each
    1 * 1 Each - 4.550,00 kr
    4.550,00 kr
    1 * 1 Each
       4.550,00 kr
  • Refill only, Oil-Only
    168,3 L
    1 * 1 Items
    1 * 1 Items - 2.950,00 kr
    2.950,00 kr
    1 * 1 Items
       2.950,00 kr
Large mobile kits: 360 litres
  • Beskrivelse
    Sugeevne
    B×D×H
  • Spill kit, Universal, includes 200 mat pads, 28 socks, 18 pillows and 15 disposal bags
    250,8 L
    850×620×1100 mm
    1 * 1 Each
    1 * 1 Each - 7.050,00 kr
    7.050,00 kr
    1 * 1 Each
       7.050,00 kr
  • Refill only, Universal
    250,8 L
    1 * 1 Items
    1 * 1 Items - 3.770,00 kr
    3.770,00 kr
    1 * 1 Items
       3.770,00 kr
  • Spill kit, HAZ-MAT, includes 160 mat pads, 23 socks, 28 pillows and 15 disposal bags
    263,5 L
    850×620×1100 mm
    1 * 1 Each
    1 * 1 Each - 8.910,00 kr
    8.910,00 kr
    1 * 1 Each
       8.910,00 kr
  • Refill only, HAZ-MAT
    263,5 L
    1 * 1 Items
    1 * 1 Items - 6.370,00 kr
    6.370,00 kr
    1 * 1 Items
       6.370,00 kr
  • Spill kit, Oil-Only, includes 160 mat pads, 14 socks, 18 pillows and 15 disposal bags
    250,8 L
    850×620×1100 mm
    1 * 1 Each
    1 * 1 Each - 8.150,00 kr
    8.150,00 kr
    1 * 1 Each
       8.150,00 kr
  • Refill only, Oil-Only
    250,8 L
    1 * 1 Items
    1 * 1 Items - 5.070,00 kr
    5.070,00 kr
    1 * 1 Items
       5.070,00 kr