Pedal Bins
Pedal bin with galvanised liner, powder-coated, hvid, 30,3 L, 292×435 mm
Catalog # 129-1892
Supplier: Rubbermaid Commercial Products
Jump to:
Product Details & Documents
These small bins are supplied with a hands-free foot pedal with a self closing lid.
- Leak-proof rigid plastic liner
- Tight-fitting lid overlaps moulded plastic gasket
- Maximum odour control and quiet operation
- Carrying handle for easy placement
Specifications
- Diameter×H:292×435 mm
- Kapacitet:30,3 L
- Farve:hvid
- Beskrivelse:Pedal bin with galvanised liner, powder-coated
- Seq:03