Orders LinkContactUsLinkComponent
Desiccator plates
Porcelain, 190 mm
Katalog # WITG8.535.005
Leverandør:   witeg Labortechnik
Desiccator plates
Katalog # WITG8.535.005
Leverandør:  witeg Labortechnik
Desiccator plates
Desiccator plates
Porcelain, 190 mm
Price
333,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Desiccator plates made from glazed porcelain. Without feet, with central hole Ø 20 mm, and many small holes Ø 5 mm.
 : DIN 12911.
Specifikationer
  • Ø:
    190 mm
  • Model:
    Porcelain