Hydrometer cylinder
500 ml, 50 mm, 350 mm
Katalog # WITG7.005.001
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Cylinder, made of PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Robust and stable
With spout and overflow vessel. This ensures that the hydrometer can be read with a completely filled cylinder.
Specifikationer
- Kapacitet:500 ml
- Indv. højde:350 mm
- Indv. Ø:50 mm