Orders LinkContactUsLinkComponent
Wipe rolls, microroll
Wipe rolls, microroll
Leverandør:   FHP VILEDA SCS
Wipe rolls, microroll
Leverandør:  FHP VILEDA SCS
Certifikater
Wipe rolls, microroll
1 tilgængelige muligheder nedenfor
Product Details & Documents
Lightweight microfibre wipe (
70% polyester, 30% polyamide) offering outstanding lint-free cleaning performance with a 99,9% removal of bacteria.
  • Good cleaning result in one step: Extremely important on delicate surfaces like glass or mirrors
  • Semi-disposable cloth-on-a-roll concept: No hygiene issues derived from cross-contamination
The ultrafine 
microfibres scrape and absorb dirt from the 
surface, leaving behind nothing but tiny water droplets. It can be used on all water resistant surfaces such as desktops, cupboards, bars, tables, as well as on sinks, toilets and tiles.
Colour: White
Product Family Options
Produktinformation
  • Farve
    L×B
    Pakket
    Tilgængelighed
    Price
  • hvid
    350×250 mm
    110 sheets/roll
    Discontinued
  • hvid
    350×250 mm
    200 sheets/roll
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 ROL
    1 * 4 ROL - 1.680,00 kr
    1.680,00 kr
    1 * 4 ROL
       1.680,00 kr
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Wall-mountable dispenser for MicroRoll and Quick 'n Dry rolls, transparent/white plastic
    Wall-mountable dispenser for MicroRoll and Quick 'n Dry rolls, transparent/white plastic
    115-1389
    Log ind for tilgængelighed
    1 * 4 Items - 1.430,00 kr