L(-)-Carnitine
Leverandør: CAMBRIDGE ISOTOPE LABS
Gå til:
Om denne vare
Synonymer: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS nummer:541-15-1
- Formel:C₇H₁₅NO₃
- MDL nummer:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
2 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS nummer:541-15-1
- Formel:C₇H₁₅NO₃
- MDL nummer:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



