Organisk referensestandard, (±)-3-Quinuclidinyl benzilate 1,000 µg/ml in acetonitrile
CAMBERQ-003
:
- Pakning:Glass bottle
- Pakn:1,2 mL
- CAS-nr:6581-06-2
- Beskrivelse:(±)-3-Quinuclidinyl benzilate 1,000 µg/ml in acetonitrile
- Formel:C₂₁H₂₃NO₃
- Beskrivelse:(±)-3-Quinuclidinyl benzilate