2-Nitrocinnamaldehyde 98%
Glass bottle, 5 g
Leverandør: Thermo Fisher Scientific
2-Nitrocinnamaldehyde 98%
Katalog # ACRO128610050
Leverandør: Thermo Fisher Scientific
CAS nummer: 1466-88-2
Gå til:
Product Details & Documents
Synonymer: 2'-Nitrocinnamaldehyde
- CAS nummer:1466-88-2
- Formel:C₉H₇NO₃
- MDL nummer:MFCD00007188
- Molecular Weight:177.16 g/mol
- Smeltende Pt:125…129 °C
- Opbevaringstemperatur:Køleskab
- EINECS:215-988-0
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to yellow-brown Powder or crystals
- Infrared spectrum:Conforms
- Melting point:125 to 129 °C
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



