Winter cap, Kara
sort, One size
Katalog # 113-1036
Leverandør: DELTA PLUS
Gå til:
Product Details & Documents
Polar fleece cap with Thinsulate® lining which keeps the head warm in low temperatures.
One size fits all.
Specifikationer
- Tøj størrelse:One size
- Farve:sort
- Seq:01
- Køn:Unisex
- Cat. No.:113-1036
- Risk area:Cold
- Model:Kara
- Outfit applikationer:General Purpose
- Stof:Polyester, Thinsulate® lining