Bandages, Askina® Elast
Elastic fixation bandage, wide-mesh, loose in the box, 4 m × 60 mm
Katalog # BRAU9048120
Leverandør: B. Braun
Gå til:
Product Details & Documents
Elastic, wide-mesh fixation bandage made from 70% viscose and 30% polyamide. For all types of fixation dressings and light support bandages.
- Very comfortable thanks to high natural fibre content
- Good adhesion of the individual turns of the bandage thanks to creped woven structure
- Approx. 130% lengthways elasticity
- Can be sterilised in an autoclave
Specifikationer
- Beskrivelse:Elastic fixation bandage, wide-mesh, loose in the box
- L×B:4 m × 60 mm