Bottle Carriers
Bottle carrier, PP, four compartments, 120×120 mm
Katalog # LPIT291187
Leverandør: LP ITALIANA
Gå til:
Product Details & Documents
Bottle carrier and container for bottles or tools, made of red PP.
- Good temperature resistance
- Good chemical resistance
- Sturdy
Specifikationer
- Beskrivelse:Bottle carrier, PP, four compartments
- Seq:02
- Rumstørrelse:120×120 mm