Orders LinkContactUsLinkComponent
Draining racks
Draining racks
400×400×600 mm
Catalog # LPIT291195
Supplier:   LP ITALIANA
Draining racks
Catalog # LPIT291195
Supplier:  LP ITALIANA
Draining racks
Draining racks
400×400×600 mm
Price
735,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Drying rack, made of PP, with 81 places that can be fully disassembled.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • With carrying handle
Specifications
  • B×D×H:
    400×400×600 mm