Pipette stand
Pipette stand, light grey polyamide, for 3 single or multi channel pipettes, 3 single or multi channel pipettes, Polyamid, lysegrå
Katalog # 635-0362
Leverandør: Socorex Isba
Gå til:
Product Details & Documents
Pipette stand made of light grey polyamide.
- For 3 single and/or multi channel pipettes
- Innovative design fits all Calibra® and Acura® models and allows easy reach of each instrument
- Heavy bottom plate and anti-slip pads guarantee stability
Specifikationer
- Til:3 single or multi channel pipettes
- Farve:lysegrå
- Beskrivelse:Pipette stand, light grey polyamide, for 3 single or multi channel pipettes
- Materiale:Polyamid