Orders LinkContactUsLinkComponent
Random hexamer primer
Random hexamer primer
Random hexamer primer 24 µg (100 µM), 1 KIT
Katalog # SO142
Leverandør:   Thermo Fisher Scientific
Random hexamer primer
Katalog # SO142
CAS nummer:  
Random hexamer primer
Random hexamer primer
Random hexamer primer 24 µg (100 µM), 1 KIT
CAS nummer:  
Price
487,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Random hexamer primer is a mixture of single-stranded random hexanucleotides with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Random hexamer primer has been functionally tested in first strand cDNA synthesis.
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Random hexamer primer 24 µg (100 µM)