Cleanroom face masks, BioClean Microflow™
Usterile, med bøjle, 375×330 mm, hvid
Katalog # 113-8158
Leverandør: Nitritex
Gå til:
Product Details & Documents
Non sterile soft face veil with headband and nosepiece manufactured using ultrasonic bonding.
- Veil made from low linting thermabond non woven material
- White hydroentangled polyester binding and tubular polyester head loop
- Fully enclosed, flexible, malleable aluminium nosepiece coated with plastic
- Processed in a cleanroom environment, contains no latex
Suitable for use in class 10, ISO Class 4, M2.5 cleanrooms.
Certifications: Manufactured in a facility holding ISO 9001:2008. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1994.
Emballering: 100 veils per double bag; 10 bags per case (1000 veils).
Documents
NITRBFV05_TDS_EN
Datablad
Specifikationer
- Model:Usterile, med bøjle
- Farve:hvid
- Størrelse:375×330 mm