Groundwater sampling capsules, Polycap™ GW, Whatman™
Product Details & Documents
These Polycap™ GW (Ground Water) capsule filters are specifically designed for sample preparation of ground water samples for dissolved metals analysis. The in-line design with stepped hose barbs and hydrophilic polyethersulfone (PES) membrane make it easy to use for field sampling.
- Stepped hose barb fittings allow for connection directly to outlet of a sampling pump with various size tubing
- Effective filtration surface area of 600 cm² to ensure rapid sample collection
- PES filters in thermally fused housing eliminate sample contamination that could come from glues, adhesives, metals, epoxies, or extraneous materials
- Lot number printed on each unit for traceability supports use in regulated field sampling methods such as EPA Method 3005 for ground water analysis
The United States Environmental Protection Agency (EPA) and other environmental protection agencies have protocols specifying support of ground water sampling with a 0,45 µm filter prior to analysing dissolved or suspended metals (EPA Method 3005).
Specifikationer
- Porestørrelse:0,45 µm
- Model:GW 75
- Seq:01
- Filtreringsareal:600 cm²
- Cat. No.:701-0164
- Maks. tryk:4,1 bar
- Indløb/udløb:6 to 9 mm stepped barb
- Flowhastighed for vand:60 l/min (1,0 bar)