Animal cell model
Animal cell model, 800 g, 210×110×310 mm
Katalog # 761-0465
Marketsource
Leverandør: 3B Scientific
Gå til:
Product Details & Documents
The two piece animal cell model shows the form and structure of a typical animal cell as viewed by an electron microscope.
For purposes of better illustration, all important organelles are shown in raised relief and displayed in colour:
Nucleus, mitochondrion, smooth endoplasmic reticulum (ER), rough endoplasmic reticulum (ER), basal membrane, collagen fibres, Golgi apparatus, microvilli, lysosome.
Specifikationer
- Vægt:800 g
- Beskrivelse:Animal cell model
- B×D×H:210×110×310 mm