Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, bent
Adapters, receiver, vacuum, bent
19/26 NS, 29/32 NS
Katalog # WITG1.906.025
Leverandør:   witeg Labortechnik
Adapters, receiver, vacuum, bent
Katalog # WITG1.906.025
Leverandør:  witeg Labortechnik
Adapters, receiver, vacuum, bent
Adapters, receiver, vacuum, bent
19/26 NS, 29/32 NS
Price
424,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3,3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Receiver adapters , bent (angle: 105°), with NS socket and NS cone, vacuum connection: glass olive, with drain tip.
Certifications: DIN 12594
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    19/26 NS