Orders LinkContactUsLinkComponent
Evaporative concentrators, Kuderna-Danish, WHEATON®
Evaporative concentrators, Kuderna-Danish, WHEATON®
Kuderna-Danish-koncentrator, komplet (270 mm synder-kolonne, kolbe, opsamlingskar og to Clear-Seal®-propper)
Catalog # WHEA297883
Supplier:   DWK Life Sciences
Evaporative concentrators, Kuderna-Danish, WHEATON®
Catalog # WHEA297883
Supplier:  DWK Life Sciences
Evaporative concentrators, Kuderna-Danish, WHEATON®
Evaporative concentrators, Kuderna-Danish, WHEATON®
Kuderna-Danish-koncentrator, komplet (270 mm synder-kolonne, kolbe, opsamlingskar og to Clear-Seal®-propper)
Price
5.120,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This unit is designed for the concentration of trace amounts of analytes in an organic solvent prior to analysis.
  • Smooth clear-seal joints eliminate the need for grease, reduces sample contamination
  • Consists of a 270 mm three-ball Snyder column, a 500 ml flask, and a 10 ml receiving vessel
  • Made of WHEATON 33 low extractable borosilicate glass meets USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
  • Beskrivelse:
    Kuderna-Danish-koncentrator, komplet (270 mm synder-kolonne, kolbe, opsamlingskar og to Clear-Seal®-propper)
  • Cat. No.:
    WHEA297883
  • Maks. kapacitet:
    500 ml
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • WHEA297825
    Kolbe, 500 ml, med 24/40 top og 19/22 bund Clear-Seal®-prop
    Log in for availability
    1 * 1 Items - 1.700,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • WHEA384353
    Synder-kolonne, 270 mm, tre kugler, med 24/20 Clear-Seal®-prop
    Log in for availability
    1 * 1 Items - 2.410,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • WHEA297814
    Kolbe, 250 ml, top 20 - 400, bund 20 - 400
    Log in for availability
    1 * 1 Items - 1.360,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • WHEA297800
    Opsamlingskar, 10 ml, 19/22 Clear-Seal®-prop, med inddeling
    Log in for availability
    1 * 1 Items - 1.080,00 kr