Leak detection additives, Reveal™
Catalog # UVPA96-0150-03
Supplier: UVP ULTRA VIOLET PRODUCTS
Specifications
- Beskrivelse:A-680 Plus-additiv til olie, brændstof og ATF-systemer
- Seq:08
- Pakn:1
Specifications
About this item
This leak detection system brings leaks to light with an ultraviolet lamp and specially formulated fluorescent tracing additives.
- Locating all leaks at once, even multiple small leaks in the same system
- Tracer additives work easily and quickly in any system using water, oil, ATF, fuel or refrigerant