Pepstatin A
Marketsource
Supplier: Serva
About this item
3 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SERA52682.02
- SERA52682.03
- SERA52682.04
- Protein/peptide name:
- Pepstatin A
- Pepstatin A
- Pepstatin A
- CAS-nr:
- 26305-03-3
- 26305-03-3
- 26305-03-3
- MDL No.:
- MFCD00060740
- MFCD00060740
- MFCD00060740
- Molecular formula:
- C₃₄H₆₃N₅O₉
- C₃₄H₆₃N₅O₉
- C₃₄H₆₃N₅O₉
- Opbevaring betingelser:
- Refrigerator
- Refrigerator
- Refrigerator
Specifications
Product Family Options
Product Information
- PakningPaknAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.SERA52682.02Protein/peptide namePepstatin ACAS-nr26305-03-3MDL No.MFCD00060740Molecular formulaC₃₄H₆₃N₅O₉Opbevaring betingelserRefrigeratorMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.SERA52682.03Protein/peptide namePepstatin ACAS-nr26305-03-3MDL No.MFCD00060740Molecular formulaC₃₄H₆₃N₅O₉Opbevaring betingelserRefrigeratorMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.SERA52682.04Protein/peptide namePepstatin ACAS-nr26305-03-3MDL No.MFCD00060740Molecular formulaC₃₄H₆₃N₅O₉Opbevaring betingelserRefrigeratorMarketsource