Carbenicillin disodium salt
Supplier: PanReac AppliChem
Jump to:
About this item
Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
- CAS Number:4800-94-6
- Formula:C₁₇H₁₆N₂O₆S·2Na
- MDL Number:MFCD00077683
- Molecular Weight:422.363 g/mol
- Storage Temperature:Køleskab
- EINECS:225-360-8
2 Options Available Below
Product Details & Documents
Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
- CAS Number:4800-94-6
- Formula:C₁₇H₁₆N₂O₆S·2Na
- MDL Number:MFCD00077683
- Molecular Weight:422.363 g/mol
- Storage Temperature:Køleskab
- EINECS:225-360-8