Streptavidin
Glass bottle for solids, 1 mg
Catalog # VECTSA-5000-1
Supplier: Vector Laboratories
Jump to:
Product Details & Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:1 mg
- Cat. No.:VECTSA-5000-1
- CAS-nr:9013-20-1
- Protein/peptide name:Streptavidin
- MDL No.:MFCD00082035
- Opbevaring betingelser:Freezer
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



