Ubbelohde viscometers for automatic measurements
Leverandør: SI Analytics
Om denne vare
Viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
The additional filling and cleaning tube and glass threads ensure safe operational use.
Certifications: ISO/DIS 3105, ASTM D2515, ASTM D446
9 tilgængelige muligheder nedenfor
Product Details & Documents
Viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
The additional filling and cleaning tube and glass threads ensure safe operational use.
Certifications: ISO/DIS 3105, ASTM D2515, ASTM D446
Specifications for Product Family
Specifikationer
- Catalog No:
- SCOI285402005
- SCOI285402013
- SCOI285402021
- SCOI285402038
- SCOI285402046
- SCOI285402054
- SCOI285402062
- SCOI285402079
- SCOI285402087
- Længde:
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- Volumen:
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
- 15 − 22 ml
Specifikationer
Product Family Options
Produktinformation
- OmrådeKonstantLængdeØ indv.VolumenTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.SCOI285402005Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402013Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402021Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402038Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402046Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402054Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402062Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402079Længde290 mmVolumen15 − 22 ml - Specifikationer:
More
Specifications
Cat. No.SCOI285402087Længde290 mmVolumen15 − 22 ml