Orders LinkContactUsLinkComponent
Special pH electrodes
Supplier:   CONSORT
Special pH electrodes
Supplier:  CONSORT
Certificates
Special pH electrodes
Special pH electrodes
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Beskrivelse
    Model
    pH
    Temperatur
    Materiale
    Elektrolyt
    Membran
    Ø×L
    Tilslutning
    Til
    Availability
    Price
  • Speciality pH electrode
    SP21X
    0 - 14
    0...+100 °C
    Glas
    Refillable
    Single junction
    12×110 mm
    S7
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Special-pH-elektrode
    SP28X, micro
    0 - 14
    0...+80 °C
    Glas
    Refillable
    Single junction
    4×130 mm
    S7
    Micro volumes (min. 10 mm depth), test tubes
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.540,00 kr
    1.540,00 kr
    1 * 1 Items
       1.540,00 kr
  • Special-pH-elektrode
    Enkelt saltbro, genopfyldelig
    0 - 14
    0...+100 °C
    Glas
    Refillable
    Single junction
    8×250 mm
    S7
    Tall form laboratory flasks, micro volumes
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.760,00 kr
    2.760,00 kr
    1 * 1 Items
       2.760,00 kr