Orders LinkContactUsLinkComponent
PIG® universal mat rolls
PIG® universal mat rolls
Leverandør:   NEW PIG
PIG® universal mat rolls
Leverandør:  NEW PIG
Certifikater
PIG® universal mat rolls
PIG® universal mat rolls
14 tilgængelige muligheder nedenfor
Product Details & Documents
Eight layers of 100% PP are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated.
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime so mat stays on the job longer
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money. Available in light, medium, or heavyweight depending on the amount of liquid you need to absorb. Light-weight construction is ideal for small tasks and cleanups. 25% or more recycled content.
Documents
Product Family Options
Produktinformation
  • Sugeevne
    Pakket
    L×B
    Tilgængelighed
    Price
  • 121-0091
    Universal mat rolls, lightweight
    Universal mat rolls, lightweight
    152,4 L
    1 rulle/pose
    91 m×760 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.450,00 kr
    2.450,00 kr
    1 * 1 Items
       2.450,00 kr
  • 121-0080
    Universal mat rolls, lightweight
    Universal mat rolls, lightweight
    76,2 L
    1 rulle/pose
    46 m×760 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.250,00 kr
    1.250,00 kr
    1 * 1 Items
       1.250,00 kr
  • 121-0095
    Universal mat rolls, lightweight
    Universal mat rolls, lightweight
    122,6 L
    1 rulle/pose
    91 m×610 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.020,00 kr
    2.020,00 kr
    1 * 1 Items
       2.020,00 kr
  • 121-0096
    Universal mat rolls, lightweight
    Universal mat rolls, lightweight
    152,4 L
    2 ruller/pose
    91 m×380 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 2.450,00 kr
    2.450,00 kr
    1 * 2 Items
       2.450,00 kr
  • 121-0092
    Universal mat rolls, lightweight
    Universal mat rolls, lightweight
    33,4 L
    2 ruller/dispenser
    31 m×250 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 909,00 kr
    909,00 kr
    1 * 2 Items
       909,00 kr
  • 121-0082
    Universal mat rolls, heavyweight
    Universal mat rolls, heavyweight
    37,1 L
    1 rulle/dispenser
    23 m×380 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 789,00 kr
    789,00 kr
    1 * 1 Items
       789,00 kr
  • 121-0533
    Universal mat rolls, mediumweight
    Universal mat rolls, mediumweight
    161 L
    1 rulle/pose
    61 m×760 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.450,00 kr
    2.450,00 kr
    1 * 1 Items
       2.450,00 kr
  • 121-0083
    Universal mat rolls, mediumweight
    Universal mat rolls, mediumweight
    120,7 L
    1 rulle/pose
    46 m×760 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.970,00 kr
    1.970,00 kr
    1 * 1 Items
       1.970,00 kr
  • 121-0085
    Universal mat rolls, mediumweight
    Universal mat rolls, mediumweight
    128 L
    1 rulle/pose
    61 m×610 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.080,00 kr
    2.080,00 kr
    1 * 1 Items
       2.080,00 kr
  • 121-0097
    Universal mat rolls, mediumweight
    Universal mat rolls, mediumweight
    161 L
    2 rolls/bag
    61 m×380 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 2.450,00 kr
    2.450,00 kr
    1 * 2 Items
       2.450,00 kr
  • 121-0522
    Universal mat rolls, heavyweight
    Universal mat rolls, heavyweight
    152,4 L
    1 rulle/pose
    46 m×760 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.450,00 kr
    2.450,00 kr
    1 * 1 Items
       2.450,00 kr
  • 121-0086
    Universal mat rolls, heavyweight
    Universal mat rolls, heavyweight
    122,6 L
    1 rulle/pose
    46 m×610 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.970,00 kr
    1.970,00 kr
    1 * 1 ROL
       1.970,00 kr
  • 121-0090
    Universal mat rolls, heavyweight
    Universal mat rolls, heavyweight
    152,4 L
    2 rulle/pose
    46 m×380 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 ROL
    1 * 2 ROL - 2.450,00 kr
    2.450,00 kr
    1 * 2 ROL
       2.450,00 kr
  • 121-0098
    Poly-back universal mat rolls
    Poly-back universal mat rolls
    77,7 L
    1 rulle/pose
    23 m×790 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 2.070,00 kr
    2.070,00 kr
    1 * 1 ROL
       2.070,00 kr
Accessories
Produktinformation
  • Katalog #
    Farve
    B×D×H
    Tilgængelighed
    Price
  • Adjustable mat and wipe roll dispenser
    Adjustable mat and wipe roll dispenser
    NEWPGEN182
    sort
    890×480×860 mm
    Log ind for tilgængelighed
    1 * 1 Items - 813,00 kr
Produktinformation
  • Katalog #
    B×D×H
    Tilgængelighed
    Price
  • Vertical mat roll holder
    Vertical mat roll holder
    NEWPGEN249
    510×510×1120 mm
    Log ind for tilgængelighed
    1 * 1 Items - 2.450,00 kr