Orders LinkContactUsLinkComponent
Methylenblåt 10 g/l in aqueous solution
Methylenblåt 10 g/l in aqueous solution
Methylenblåt 10 g/l in aqueous solution
Supplier:  VWR Chemicals
Certificates
Methylenblåt 10 g/l in aqueous solution
2 Options Available Below
Product Details & Documents
Synonyms:  Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
  • CAS Number:61-73-4
  • Formula:C₁₆H₁₈ClN₃S
  • MDL Number:MFCD00150008
  • Molecular Weight:319.86 g/mol
  • Density:1,01 g/cm³ (20 °C)
  • EINECS:200-515-2
Specifications for Product Family
Specification Test Results
  • Identification:
    Passes test
  • Control weight:
    QC checked
Product Family Options
Product Information