Orders LinkContactUsLinkComponent
Standard storage cabinets, Protector®
Standard storage cabinets, Protector®
Leverandør:   LABCONCO
Standard storage cabinets, Protector®
Leverandør:  LABCONCO
Certifikater
Standard storage cabinets, Protector®
Standard storage cabinets, Protector®
7 tilgængelige muligheder nedenfor
Product Details & Documents
Storage cabinets support fume hoods and used for storage and venting of small laboratory instruments, mild acids and other reagents.
  • Manual-closing, non-locking doors
  • Durable epoxy coated steel construction
Leveringsinformation: Includes four leveling feet and one filter panel (20 cm) to increase cabinet depth from 55 to 76,2 cm. Spillstopper™ work surfaces, flat work surfaces, shelf kits and filter panels can be ordered separately.
Product Family Options
Produktinformation
  • Døre
    B×D×H
    Vægt
    Tilgængelighed
    Price
  • LBCP9900300
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    Singler right hinge
    610×560×900 mm
    59 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • LBCP9900200
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    2
    760×560×900 mm
    70 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • LBCP9900100
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    2
    910×560×900 mm
    82 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • LBCP9900000
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
    2
    1220×560×900 mm
    118 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • LBCP9900400
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
    Single, right hinge
    457×559×900 mm
    41 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • LBCP9900600
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
    Single, left hinge
    610×559×900 mm
    59 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • LBCP9900700
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
    Single, left hinge
    457×559×900 mm
    41 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Interior epoxy coated shelf and mounting hardware, 762 mm width, 4,5 kg
    Interior epoxy coated shelf and mounting hardware, 762 mm width, 4,5 kg
    LBCP9919102