Single-channel pipettes, electronic, variable volume, Picus®
Catalog # 613-1967
Supplier: Sartorius
Specifications
- Volumen:10 - 300 µl
- Seq:03
- Environmentally Preferable:Lav fremstillingspåvirkning
- Miljøcertificering:Renewable energy certificate; ISO 14001
- Forøgelse:0,2 µl
- Accuracy:±0,40 %
- Imprecision:<0,15 %
- Pakn:1
- Recommended tips:350 / F300 µl
- Med Safe-Cone filtre:613-4070, 613-0693
Specifications
About this item
Small, lightweight electronic pipettes that eases the user's workload, protects user from injury by ensuring optimal working posture and allows accurate and precise pipetting results.
- Helps to avoid pipetting-related symptoms like RSI (repetitive strain injury)
- High ergonomic standards with electronic tip ejection, adjustment wheel for scrolling menus, adjusting volume and controlling manual pipetting and feather-light operating button
- Error control system guarantees result reliability
- Unique tracker in conjunction with pipetting, reverse pipetting and multi-dispensing modes to help microplate work
- Easy maintenance of lower parts with GLP information integrated in software for service and calibration follow-up
- Autoclavable (lower parts only)
There are two types of filters: Safe-cone standard filter and safe-cone plus filter. Standard filters are recommended for general use while the plus filter is for more demanding applications, such as cell culture, bacterial and viral work plus molecular biology.
Ordering information: Supplied with USB cable. Charging stand and USB adapter available separately.