Stirrers tools, adjustable rotor, PTFE
Leverandør: COWIE
Gå til:
Om denne vare
Adjustable rotors made of PTFE with glass filled PTFE lock nut.
- Resistant to chemicals
- Can be use up to 280 °C
These rotors slide over standard shafts and lock in place to give the most effective stirring patterns.
25 tilgængelige muligheder nedenfor
Product Details & Documents
Adjustable rotors made of PTFE with glass filled PTFE lock nut.
- Resistant to chemicals
- Can be use up to 280 °C
These rotors slide over standard shafts and lock in place to give the most effective stirring patterns.
Product Family Options
Angled type 45°, 4 blade
Produktinformation
- Omrører - ØSkaft - ØTilgængelighedPrice
- COWI005.608040Angled type 45° metric
- COWI005.610060Angled type 45° metric
- COWI005.612070Angled type 45° metric
- COWI005.612090Angled type 45° metric
- COWI005.6.250.15Angled type 45° imperial
- COWI005.616100Angled type 45° metric
- COWI005.606040Angled type 45° metric
- COWI005.6.625.40Angled type 45° imperial
- COWI005.6.500.35Angled type 45° imperial
- COWI005.6.375.25Angled type 45° imperial
- COWI005.6.313.15Angled type 45° imperial
- COWI005.610090Angled type 45° metric
Flat type
Produktinformation
- Omrører - ØSkaft - ØTilgængelighedPrice
- COWI005.708070Flat type, metric
- COWI005.710070Flat type, metric
- COWI005.710100Flat type, metric
- COWI005.712100Flat type, metric
- COWI005.712150Flat type, metric
- COWI005.7.313.3Flat type, imperial
- COWI005.716150Flat type, metric
- COWI005.716100Flat type, metric
- COWI005.706070Flat type, metric
- COWI005.7.625.6Flat type, imperial
- COWI005.7.375.3Flat type, imperial
- COWI005.7.250.3Flat type, imperial
- COWI005.7.500.4Flat type, imperial