Stopcocks, needle-valve, angled
0 - 6 mm
Katalog # WITG0.243.250
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Needle valve stopcock, Duran, with PTFE-needle valve, sealing system and overtwist safety device.
- Very good chemical resistance
- Very good temperature resistance
Specifikationer
- Hul Ø:0 - 6 mm