Rain jacket, Morgat 4391
Om denne vare
Water- and windproof jacket made of Flexothane® KLEEN: 100% polyamide 6.6 knitting with PU coating. With taped seams. Washable at 95 °C. Ideal for food applications.
- Supple, stretchable, tear-resistant and comfortable
- Fixed hood
- Zip closure under flap with press studs
- Raglan sleeves
- Elasticated wristband
- Underarm ventilation
Weight: 170 g/m²
Certifications: ENV 343 (3-1); EN 14605 (Type PB [4])
Food contact approved
12 tilgængelige muligheder nedenfor
Product Details & Documents
Water- and windproof jacket made of Flexothane® KLEEN: 100% polyamide 6.6 knitting with PU coating. With taped seams. Washable at 95 °C. Ideal for food applications.
- Supple, stretchable, tear-resistant and comfortable
- Fixed hood
- Zip closure under flap with press studs
- Raglan sleeves
- Elasticated wristband
- Underarm ventilation
Weight: 170 g/m²
Certifications: ENV 343 (3-1); EN 14605 (Type PB [4])
Food contact approved
Specifications for Product Family
Specifikationer
- Catalog No:
- SIOE4391/W02/S
- SIOE4391/W02/M
- SIOE4391/W02/L
- SIOE4391/W02/1XL
- SIOE4391/W02/2XL
- SIOE4391/W02/3XL
- 113-2217
- 113-2218
- 113-2219
- 113-2220
- 113-2221
- 113-2222
- Model:
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- 4391 Morgat
- Outfit applikationer:
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Food|Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- 100% polyamide 6.6 knitting with PU coating
- Vægt:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.SIOE4391/W02/SModel4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE4391/W02/MModel4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE4391/W02/LModel4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE4391/W02/1XLModel4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE4391/W02/2XLModel4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE4391/W02/3XLModel4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.113-2217Model4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.113-2218Model4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.113-2219Model4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.113-2220Model4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.113-2221Model4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.113-2222Model4391 MorgatOutfit applikationerFood|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyamide 6.6 knitting with PU coatingVægt170 g/m²