Fleece jacket, DYNAMIC® Tortolas 443
Leverandør: SIOEN
Om denne vare
Single-coloured jacket for men, made of 2-layer bonded polyester fleece. Both sides are brushed, one side features an anti-pilling treatment. Can be zipped into different jackets.
- Straight collar, hidden zip closure
- Inset chest pocket with zip closure, two inset pockets with zip closure
- Round inset sleeves, elastic piping at end of sleeves
- Drawstring in hem
Weight: 320 g/m²
Certifications: EN 340 (1XXXX)
6 tilgængelige muligheder nedenfor
Product Details & Documents
Single-coloured jacket for men, made of 2-layer bonded polyester fleece. Both sides are brushed, one side features an anti-pilling treatment. Can be zipped into different jackets.
- Straight collar, hidden zip closure
- Inset chest pocket with zip closure, two inset pockets with zip closure
- Round inset sleeves, elastic piping at end of sleeves
- Drawstring in hem
Weight: 320 g/m²
Certifications: EN 340 (1XXXX)
Specifications for Product Family
Specifikationer
- Catalog No:
- SIOE443Z/B95/S
- SIOE443Z/B95/M
- SIOE443Z/B95/L
- SIOE443Z/B95/1XL
- SIOE443Z/B95/2XL
- SIOE443Z/B95/3XL
- Model:
- Tortolas 443
- Tortolas 443
- Tortolas 443
- Tortolas 443
- Tortolas 443
- Tortolas 443
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Køn:
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Vægt:
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.SIOE443Z/B95/SModelTortolas 443Outfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStofPolyesterVægt320 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE443Z/B95/MModelTortolas 443Outfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStofPolyesterVægt320 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE443Z/B95/LModelTortolas 443Outfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStofPolyesterVægt320 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE443Z/B95/1XLModelTortolas 443Outfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStofPolyesterVægt320 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE443Z/B95/2XLModelTortolas 443Outfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStofPolyesterVægt320 g/m² - Specifikationer:
More
Specifications
Cat. No.SIOE443Z/B95/3XLModelTortolas 443Outfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStofPolyesterVægt320 g/m²