Jacket, Pro Industry
Jacket made of 65% polyester and 35% cotton, ideal for light-duty industrial applications. Made for the industrial worker with emphasis on comfort, durability and functionality.
- Adjustable waist and cuffs
- Two front pockets, two chest pockets with flap, one with pen pocket
- Inside pocket, inside mobile phone pocket with velcro
Weight: 245 g/m²
Jacket made of 65% polyester and 35% cotton, ideal for light-duty industrial applications. Made for the industrial worker with emphasis on comfort, durability and functionality.
- Adjustable waist and cuffs
- Two front pockets, two chest pockets with flap, one with pen pocket
- Inside pocket, inside mobile phone pocket with velcro
Weight: 245 g/m²
Documents
- Catalog No:
- 113-4104
- 113-4103
- 113-4102
- 113-4101
- 113-4099
- 113-4100
- 113-4122
- 113-4121
- 113-4120
- 113-4119
- 113-4117
- 113-4118
- 113-4116
- 113-4115
- 113-4114
- 113-4113
- 113-4111
- 113-4112
- Model:
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Pro Industry
- Outfit applikationer:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Køn:
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Stof:
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- Vægt:
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
Product Family Options
- FarveTøj størrelse
Specifications
Cat. No.113-4104ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4103ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4102ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4101ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4099ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4100ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4122ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4121ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4120ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4119ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4117ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4118ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4116ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4115ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4114ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4113ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4111ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²Specifications
Cat. No.113-4112ModelPro IndustryOutfit applikationerIndustrialKønFor mændSleeve or leg lengthLongLukkestørrelseMed Velcro®Stof65% polyester and 35% cottonVægt245 g/m²