Softshell jacket, Piemonte 9834
: SIOEN
Lightweight, modern softshell jacket for protection against the cold. The jacket is made of two-layer bonded softshell (100% polyester stretch) on the outside and 100% polyester fleece on the inside.
- Fixed windbreaker lining in front
- Straight collar, zip closure with inside flap
- One inset chest pocket and two standard inset pockets with zip closure
- Raglan ergonomic sleeve, sleeve narrowing by touch and close fastening
Weight: 250 g/m²
Certifications: EN 14058: 2004 (1XXXX)
Lightweight, modern softshell jacket for protection against the cold. The jacket is made of two-layer bonded softshell (100% polyester stretch) on the outside and 100% polyester fleece on the inside.
- Fixed windbreaker lining in front
- Straight collar, zip closure with inside flap
- One inset chest pocket and two standard inset pockets with zip closure
- Raglan ergonomic sleeve, sleeve narrowing by touch and close fastening
Weight: 250 g/m²
Certifications: EN 14058: 2004 (1XXXX)
- Catalog No:
- 113-08246
- 113-08247
- 113-08248
- 113-08249
- 113-08250
- 113-08251
- 113-08258
- 113-08259
- 113-08260
- 113-08261
- 113-08262
- 113-08263
- 113-08264
- 113-08265
- 113-08266
- 113-08267
- 113-08268
- 113-08269
- Model:
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- 9834 Piemonte
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Køn:
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- Vægt:
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
Product Family Options
- FarveTøj størrelse
Specifications
Cat. No.113-08246Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08247Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08248Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08249Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08250Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08251Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08258Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08259Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08260Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08261Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08262Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08263Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08264Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08265Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08266Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08267Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08268Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²Specifications
Cat. No.113-08269Model9834 PiemonteOutfit applikationerOutdoorRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyesterVægt250 g/m²