Orders LinkContactUsLinkComponent
Electrical resistant gloves, Electrosoft CL 0
Electrical resistant gloves, Electrosoft CL 0
Supplier:   HONEYWELL SAFETY
Electrical resistant gloves, Electrosoft CL 0
Supplier:  HONEYWELL SAFETY
Certificates
Electrical resistant gloves, Electrosoft CL 0
2 Options Available Below
Product Details & Documents
Natural latex insulating gloves for electrical work with a maximum voltage of 1000 volts.
  • Ergonomic shape and lightly powdered interior make the glove easy to slip on and off
  • Natural latex base offers high dielectric characteristics
  • Thickness ensures good dexterity while offering protection up to 1000 volts
Please note: Must be used with a leather overglove.
Thickness: 1 mm
Colour: Beige
EN 60903-2003, CEI 903-2002; Cat. III
Certifications: EN 60903-2003, CEI 903-2002; Cat. III
Specifications for Product Family
Specifications
  • Catalog No:
  • DALL2091907/8
  • DALL2091907/11
  • Model:
  • CL 0
  • CL 0
  • Ambidextrous/Hånd-specifik:
  • Hånd-specifik
  • Hånd-specifik
  • Engangs/Genbrugelig:
  • Genanvendelig
  • Genanvendelig
  • Med pudder/pudderfri:
  • Lav pudder
  • Lav pudder
  • Overflade:
  • Smooth
  • Smooth
  • Manchet:
  • Lang tyk
  • Lang tyk
  • EN norm:
  • EN 60903-2003, CEI 903-2002
  • EN 60903-2003, CEI 903-2002
Specifications
Product Family Options
Product Information
  • Handske størrelse
    Farve
    Længde
    Tykkelse
    Availability
    Price
  • 8
    beige
    360 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 1 Par
    1 * 1 Par
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2091907/8
    Model
    CL 0
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Med pudder/pudderfri
    Lav pudder
    Overflade
    Smooth
    Manchet
    Lang tyk
    EN norm
    EN 60903-2003, CEI 903-2002
  • 11
    beige
    360 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 1 Par
    1 * 1 Par
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2091907/11
    Model
    CL 0
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Med pudder/pudderfri
    Lav pudder
    Overflade
    Smooth
    Manchet
    Lang tyk
    EN norm
    EN 60903-2003, CEI 903-2002
Leather overgloves
Product Information
  • Catalog #
    Farve
    Tykkelse
    Handske størrelse
    Længde
    Availability
    Price
  • grå/beige
    0,8 - 1,5 mm
    10
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 Par
    1 * 1 Par - 0,00 kr
    0,00 kr
    1 * 1 Par
       0,00 kr
Product Information
  • Catalog #
    Farve
    Tykkelse
    Handske størrelse
    Længde
    Availability
    Price
  • grå/beige
    0,8 - 1,5 mm
    9
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 Par
    1 * 1 Par - 0,00 kr
    0,00 kr
    1 * 1 Par
       0,00 kr